missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix M, 4.5mL Tube

MP Biomedicals™ Lysing Matrix M, 4.5mL Tube allows for lysing of any kind of difficult to homogenize samples for DNA, RNA and Protein Extractions using the BigPrep™ adapter

$691.88 - $2,075.78

Specifications

Product Type Lysis Matrix
For Use With (Application) Sample preparation, Sample lysis, Nucleic acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis; Gene function, Gene regulation
For Use With (Equipment) FastPrep-24, FastPrep 24 5G, FastPrep-96
Matrix M
Storage Requirements Room temperature
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP116949025 View Documents MP Biomedicals™
116949025
25 x 4.5mL Each for $691.88
 
MP116949100 View Documents MP Biomedicals™
116949100
100 x 4.5mL Each for $2,075.78
 
Description

Description

  • Universal grinding
  • Homogenization and Lysing Matrix
  • Lyse any kind of difficult to homogenize samples for DNA, RNA and protein extractions using the BigPrep™ adapter

Certifications

DNase and Rnase free

Specifications

Specifications

Lysis Matrix
FastPrep-24, FastPrep 24 5G, FastPrep-96
Room temperature
Sample preparation, Sample lysis, Nucleic acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis; Gene function, Gene regulation
M