missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix C Bulk

Manufacturer:  MP Biomedicals™ 116540433

 View more versions of this product

Catalog No. MP116540433



Description

Description

  • 1mm silica spheres
  • Used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae
  • Found in the FastRNA Pro Red Kit and FastProtein Red Matrix
Specifications

Specifications

Lysing Matrix C
1kg
Store at 2° - 8°C
Documents