missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix B Bulk

Used primarily for lysis of gram positive and gram negative bacteria, can also be used as an additive for disruption by sonication.

Manufacturer:  MP Biomedicals™ 116540429

 View more versions of this product

Catalog No. MP116540429



Description

Description

  • Component of FastPrep™ sample preparation system
  • Found in FastRNA™ Pro Blue Kit and FastProtein™ Blue Matrix
Specifications

Specifications

Lysis of bacteria
White Glass Beads (Silica); Single Tubes
Bacteria (Gram positive and negative); Spores
0.074 to 0.150mm
FastPrep-24; FastPrep 120; FastPrep-9 homogenizer
1kg
Store at room temperature (15 to 30°C)
Documents