Please login to your online account to display your discounted pricing

Alfa Aesar™ [Lys&superscript_1;, Pro&superscript_2;,&superscript_5;, Arg&superscript_3;,&superscript_4;, Tyr&superscript_6;] Vasoactive Intestinal Peptide, Human, Porcine, Rat

For cellular and molecular biology applications

Specifications

Product Type [Lys1, Pro2,5, Arg3,4, Tyr6] Vasoactive Intestinal Peptide, human, porcine, rat
Cross Reactivity Human, Porcine, Rat,
Molecular Weight 3467.10
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66794EXD

 
alfa aesar™
J66794EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66794MCR

 
alfa aesar™
J66794MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Lys1, Pro2,5, Arg3,4, Tyr6] Vasoactive Intestinal Peptide, human, porcine, rat
Cross Reactivity Human, Porcine, Rat,
Molecular Weight 3467.10
Storage Requirements -30° to -10°C
Format Solid
Sequence Lys-Pro-Arg-Arg-Pro-Tyr-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
Common Name [Lys1, Pro25, Arg34, Tyr6] VIP, human, porcine, rat
Regulatory Status RUO
Purification Method HPLC