Alfa Aesar™ [Lys&superscript_1;, Pro&superscript_2;,&superscript_5;, Arg&superscript_3;,&superscript_4;, Tyr&superscript_6;] Vasoactive Intestinal Peptide, Human, Porcine, Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66794EXD
 View more versions of this product
Catalog No. AAJ66794EXD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
3467.10
Human, Porcine, Rat,
Solid
RUO
[Lys1, Pro25, Arg34, Tyr6] VIP, human, porcine, rat
HPLC
0.5mg
Lys-Pro-Arg-Arg-Pro-Tyr-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
-30° to -10°C
[Lys1, Pro2,5, Arg3,4, Tyr6] Vasoactive Intestinal Peptide, human, porcine, rat