Please login to your online account to display your discounted pricing

Alfa Aesar™ Lys-Gly-Asp-Ser

For cellular and molecular biology applications

Specifications

Product Type Lys-Gly-Asp-Ser
Molecular Weight 405.40
Storage Requirements -30° to -10°C
Format Solid
Sequence K-G-D-S
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66843MA

 
alfa aesar™
J66843MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66843MB

 
alfa aesar™
J66843MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Lys-Gly-Asp-Ser
Molecular Weight 405.40
Storage Requirements -30° to -10°C
Format Solid
Sequence K-G-D-S
Regulatory Status RUO
Purification Method HPLC