Please login to your online account to display your discounted pricing

Alfa Aesar™ [Lys&superscript_3;] Bombesin


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66697MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66697MCR

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Quantity 1mg
Molecular Weight 1591.83
Format Solid
Product Type [Lys3] Bombesin
Sequence Glp-Gln-Lys-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Regulatory Status RUO
Purification Method HPLC