Please login to your online account to display your discounted pricing

Alfa Aesar™ [Lys&superscript_0;-Ala&superscript_3;] Bradykinin

For cellular and molecular biology applications

Specifications

Product Type [Lys0-Ala3] Bradykinin
Molecular Weight 1162.34
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66738LB0

 
alfa aesar™
J66738LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66738MB

 
alfa aesar™
J66738MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Lys0-Ala3] Bradykinin
Molecular Weight 1162.34
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC