Please login to your online account to display your discounted pricing

Alfa Aesar™ [Lys&superscript_0;-Ala&superscript_3;] Bradykinin


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66738LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66738LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Storage Requirements -30° to -10°C
Regulatory Status RUO
Sequence Lys-Arg-Pro-Ala-Gly-Phe-Ser-Pro-Phe-Arg
Purification Method HPLC
Product Type [Lys0-Ala3] Bradykinin
Molecular Weight 1162.34