missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Human Lyn Isoform B, His Tag Recombinant Protein
Supplier: Thermo Scientific™ P2907
Description
LYN is a SRC family kinase expressed predominantly in hematopoietic cells. Activation of LYN has been shown to lead to imatinib resistance in CML patients. LYNB and the slightly shorter LYNA are the two isoforms of LYN.Specifications
AAH59394 | |
4067 | |
10 μg | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 0.05 mM EDTA, 0.05% NP-40, 1 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
10 μg | |
Dry Ice | |
C-Terminal | |
v-yes-1 Yamaguchi sarcoma viral related oncogene homolog | |
Kinases & Inhibitors | |
Cytoplasmic Tyrosine Kinases |
Kinase Assay | |
57.4 kDa | |
Lyn B | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
LYNB | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Oncology, Signal Transduction |