missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ LYN A Recombinant Human Protein, N-GST tag DFS Item

LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog) is a SRC family kinase expressed predominantly in hematopoietic, neural, liver, and adipose tissue.

$608.00 - $8,640.00

Specifications

Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6448 View Documents Invitrogen™
LSPV6448
10μg Each for $608.00
 
PV6449 View Documents Invitrogen™
LSPV6449
100μg Each for $2,000.00
 
PV6450 View Documents Invitrogen™
LSPV6450
1mg Each for $8,640.00
 
Description

Description

LYN is a protein-coding gene and is associated with diseases such as alexia and chorea-acanthocytosis. Activation of LYN has been shown to lead to imatinib resistance in CML patients. LYN B and the slightly shorter LYN A are the two isoforms of LYN. An important paralog of this gene is FRK. The full length LYN A protein enables researchers to study important mechanisms underlying disease.

  • Increase disease relevance by interrogating a purified and active full-length LYN A reagent
  • Uncover novel compounds or hits specific to the full length form
  • Advance your research using the full-length protein and validated assays
  • Assay Platforms: LanthaScreen™ TR-FRET Kinase Activity Assay, LanthaScreen™ Eu Kinase Binding Assay, and Z’-LYTE™ Kinase Activity Assay
Specifications

Specifications

Human
GST Tag
Yes