Please login to your online account to display your discounted pricing

Lymphotoxin beta Receptor Rat anti-Mouse, Biotin, Clone: eBio3C8 (3C8), eBioscience™

Rat Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen Lymphotoxin beta Receptor
Clone eBio3C8 (3C8)
Host Species Rat
Gene Alias LTbR,, LTbeta R
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011899

 
life technologies
LS13567180
25μg Each for $158.05

5011900

 
life technologies
LS13567182
100μg Each for $404.55
Description & Specifications

Specifications

Antigen Lymphotoxin beta Receptor
Clone eBio3C8 (3C8)
Host Species Rat
Gene Alias LTbR,, LTbeta R
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The antibody reacts with mouse lymphotoxin-beta receptor (LTBR). Northern blot analysis of tissues from adult mice showed that expression levels of LTBR mRNA were strong in lung, liver, and kidney, moderate in heart and testes, but weak in brain, thymus, spleen, and lymph nodes. The tumor necrosis factor receptor-related protein is the human receptor for the heterotrimer of lymphotoxin-alpha and lymphotoxin-beta. This LT-alpha/LT-beta heterotrimer (LT-α1β2) is assumed to take part in immunologic reactions by cell-cell contact, but does not bind to either TNFR1 (CD120α) or TNFR2 (CD120β). LTBR is expressed by day 7 embryos, so it has been speculated that the LT-alpha/LT-beta receptor system may also have some function in early embryogenesis.