Please login to your online account to display your discounted pricing

Lyle and Louise: Bad Impressions™ Bite Marks Analysis

Introduces forensic odontology and the techniques used for bite mark analysis.

$187.80 - $187.80

Specifications

Class Size 30 students
Grade Level High school and undergraduate courses
Includes 6 photos of bite mark, 60 pieces of base plate wax, 12 pieces of evidence wax, 6 evidence labels, 6 protractors, teaching manual
Storage Requirements Room temperature
View More Specs
Includes:  Six photos of bite marks, 60 pieces of base plate wax, 12 pieces of evidence wax, six evidence labels, six protractors, teaching manual
Products
Catalog Number Mfr. No. Requires For Use With (Application) Product Type Price Quantity    

S94588

 
fisher science education
VXH10110
Zipper bags (optional), lab gloves (optional) For teaching bite mark analysis procedures Mystery Of Lyle And Louise: Bad Impressions™ Bite Marks Analysis Forensic Science Curriculum Kit Each for $187.80
Description & Specifications

Specifications

Class Size 30 students
Grade Level High school and undergraduate courses
Includes 6 photos of bite mark, 60 pieces of base plate wax, 12 pieces of evidence wax, 6 evidence labels, 6 protractors, teaching manual
Storage Requirements Room temperature

A bite mark found on John Wayne Gretzky's arm during the car crash investigation casts suspicion of wrongdoing. Students conduct bite mark analysis to determine if the bite mark comes from a murder victim.

  • Students take casts of their own teeth in soft wax and identify their bite pattern
  • Using the techniques presented, they develop their own personal set of objective criteria for bite mark analysis
  • With these criteria, they compare wax casts of the victims with a photo of the bite mark on John Wayne's arm to determine if it could have been made by any of the victims
  • Class size: six groups of up to five students per group
  • For high school and undergraduate courses
  • Meets National Science Education Standards