Please login to your online account to display your discounted pricing

LY 78335, Tocris™

Phenylethanolamine-N-methyltransferase inhibitor

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material LY 78335
Assay Percent Range >99%
Molecular Formula C8H9Cl2N·HCl
Formula Weight 226.53
Synonym 2, 3-Dichloro-α-methylbenzylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

406050

 
r & d systems
RDS406050
50mg Each for $567.83

406010

 
r & d systems
RDS406010
10mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 226.53
Quantity 50mg
Chemical Name or Material LY 78335
Assay Percent Range >99%
Molecular Formula C8H9Cl2N·HCl
CAS 39959-66-5
Synonym 2, 3-Dichloro-α-methylbenzylamine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature