missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-6G (Gr-1) Rat anti-Mouse, PerCP-eFluor 710, Clone: 1A8-Ly6g, eBioscience™

Rat Monoclonal Antibody

$145.00 - $442.25

Specifications

Antigen Ly-6G (Gr-1)
Clone 1A8-Ly6g
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123955 Life Technologies
LS46966880
25μg Each for $145.00
501123956 Life Technologies
LS46966882
100μg Each for $442.25
Specifications

Description

The monoclonal antibody 1A8-Ly6g reacts with mouse Ly-6G, which is also known as Gr-1. Ly-6G is a 25kDa GPI protein expressed exclusively by neutrophils. Studies suggest an important role in neutrophil infiltration, recruitment and migration. Ly-6G associates with B2 integrins CD11a and CD11b and can attenuate B2 integrin expression and function. The ligand for Ly-6G is currently unknown.

Unlike the RB6-8C5 antibody, 1A8-Ly6g is reported to only bind to Ly-6G found on neutrophils. Moreover, 1A8-Ly6g has been used to deplete neutrophils and reported to have antagonist activity.
Description & Specifications

Specifications

Ly-6G (Gr-1)
Rat
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
1A8-Ly6g
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer