Please login to your online account to display your discounted pricing

Ly-6G (Gr-1) Rat anti-Mouse, PE-Cy5, Clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen Ly-6G (Gr-1)
Clone RB6-8C5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014147

 
life technologies
LS15593181
50μg Each for $216.05

5014149

 
life technologies
LS15593183
200μg Each for $506.05

501128871

 
life technologies
LS15593182
100μg Each for $368.30
Description & Specifications

Specifications

Antigen Ly-6G (Gr-1)
Clone RB6-8C5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.