missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-6G (Gr-1) Rat anti-Mouse, eFluor 660, Clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

$157.08 - $302.28

Specifications

Antigen Ly-6G (Gr-1)
Clone RB6-8C5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124411 Life Technologies
LS50593182
100μg Each for $302.28
5016459 Life Technologies
LS50593180
25μg Each for $157.08
Specifications

Description

The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.
Description & Specifications

Specifications

Ly-6G (Gr-1)
Rat
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
RB6-8C5
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer