missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-6G (Gr-1) Rat anti-Mouse, Clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

$71.05 - $578.55

Specifications

Antigen Ly-6G (Gr-1)
Clone RB6-8C5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry,Functional Assay,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122702 Life Technologies
LS14593186
1mg Each for $578.55
5013329 Life Technologies
LS14593181
50μg Each for $71.05
5013330 Life Technologies
LS14593182
100μg Each for $114.55
5013331 Life Technologies
LS14593185
500μg Each for $259.55
Specifications

Description

The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.
Description & Specifications

Specifications

Ly-6G (Gr-1)
Rat
Flow Cytometry,Functional Assay,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
RB6-8C5
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer