Please login to your online account to display your discounted pricing

Ly-6C Rat anti-Mouse, PE, Clone: HK1.4, eBioscience™

Rat Monoclonal Antibody

$162.40 - $390.05

Specifications

Antigen Ly-6C
Clone HK1.4
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245507

 
life technologies
LS12593280
25μg Each for $162.40

50245508

 
life technologies
LS12593282
100μg Each for $390.05
Description & Specifications

Specifications

Antigen Ly-6C
Clone HK1.4
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody HK1.4 recognizes mouse Ly-6C, a GPI-linked protein of the Ly6 family. Ly-6C is found on monocytes/macrophages, endothelial cells and granulocytes as well as a subset of lymphocytes. Some variation of expression is found on different mouse strains in regards to expression on CD4 and CD8 lymphocytes. These correlate to 2 alleles both of which are recognized by HK1.4: Ly6c.1 found on C57Bl/6 and SJL cells which results in staining of both CD4 and CD8 cells while Ly6-C.2 found on BALB/c and 3H/He results in staining of CD8, but not CD4 cells.

In vitro addition with HK1.4 antibody can increase proliferation and stimulate cytokine release.