missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-6C, APC-eFluor 780, clone: HK1.4, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS47593282

 View more versions of this product

Catalog No. 501129287



Description

Description

Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications

Specifications

Ly-6C
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P0CW02
AA682074, AA959465, Ly-6C, Ly6c, Ly6c2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
HK1.4
APC-eFluor® 780
Ly6c1
Liquid
Ly6c1
IgG2c, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17067
Documents

For Research Use Only.