Please login to your online account to display your discounted pricing

Ly-6A/E (Sca-1) Rat anti-Mouse, PE-Cy5, Clone: D7, eBioscience™

Rat Monoclonal Antibody

$265.35 - $501.70

Specifications

Antigen Ly-6A/E (Sca-1)
Clone D7
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014153

 
life technologies
LS15598181
50μg Each for $265.35

5014154

 
life technologies
LS15598182
100μg Each for $501.70
Description & Specifications

Specifications

Antigen Ly-6A/E (Sca-1)
Clone D7
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The D7 monoclonal antibody reacts with mouse Sca-1, an 18kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset.