missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-6A/E (Sca-1) Rat anti-Mouse, PE, Clone: D7, eBioscience™

Rat Monoclonal Antibody

$145.20 - $368.28

Specifications

Antigen Ly-6A/E (Sca-1)
Clone D7
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011049 Life Technologies
LS12598182
100μg Each for $269.28
5011048 Life Technologies
LS12598181
50μg Each for $145.20
5011050 Life Technologies
LS12598183
200μg Each for $368.28
Specifications

Description

The D7 monoclonal antibody reacts with mouse Sca-1, an 18kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset.
Description & Specifications

Specifications

Ly-6A/E (Sca-1)
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
D7
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL