missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-49I Mouse anti-Mouse, PE, Clone: YLI-90, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $259.55

Specifications

Antigen Ly-49I
Clone YLI-90
Host Species Murine
Gene Alias Ly49I
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010968 Life Technologies
LS12589581
50μg Each for $158.05
5010969 Life Technologies
LS12589582
100μg Each for $259.55
Specifications

Description

The YLI-90 monoclonal antibody reacts with the B6 mouse Ly-49I receptor. Ly49I, a member of the Ly49 family with inhibitory function, is a homodimer type II transmembrane protein and contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM). YLI-90 binds to subpopulations of NK, NKT and T cell lineages in the C57Bl/6 strain. This antibody does not bind to cells from BALB/c, SJL or AKR/J mice.
Description & Specifications

Specifications

Ly-49I
Murine
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
YLI-90
Ly49I
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL