missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-49H Mouse anti-Mouse, Clone: 3D10, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $170.28

Specifications

Antigen Ly-49H
Clone 3D10
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry,Functional Assay,Immunoprecipitation
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013276 Life Technologies
LS14588681
50μg Each for $114.55
5013277 Life Technologies
LS14588682
100μg Each for $170.28
Specifications

Description

The monoclonal antibody 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D (cat 12-5783). Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells.
Description & Specifications

Specifications

Ly-49H
Murine
Flow Cytometry,Functional Assay,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
3D10
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer