Please login to your online account to display your discounted pricing

Ly-49C/I/F/H Golden Syrian Hamster anti-Mouse, PE, Clone: 14B11, eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$129.05 - $245.05

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Host Species Golden Syrian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011070

 
affymetrix
12599182
100μg Each for $245.05

5011069

 
affymetrix
12599181
50μg Each for $129.05
Description & Specifications

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Host Species Golden Syrian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 14B11 monoclonal antibody reacts with a subset of mouse Ly-49 receptors and recognizes a majority of mouse NK cells. This antibody reacts with 3 putative inhibitory members of the Ly-49 family, F, I, and C and one putative activating receptor, Ly-49H. 14B11 does not react with Ly-49A, B, D, or G. It is reported that 14B11 enhances the redirected lysis of FcR+ target cells by IL-2-activated NK cells.