Please login to your online account to display your discounted pricing

LY 487379 hydrochloride, Tocris

Positive allosteric modulator selective for mGlu2

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material LY 487379 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H19F3N2O4S·HCl
Formula Weight 488.91
Synonym 2, 2, 2-Trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(3-pyridinylmethyl)ethanesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

328350

 
r & d systems
RDS328350
50mg Each for $862.95

328310

 
r & d systems
RDS328310
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 488.91
Quantity 50mg
Assay Percent Range >99%
Molecular Formula C21H19F3N2O4S·HCl
CAS 353229-59-1
Chemical Name or Material LY 487379 hydrochloride
Synonym 2, 2, 2-Trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(3-pyridinylmethyl)ethanesulfonamide hydrochloride
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C