Please login to your online account to display your discounted pricing

LY 456236 hydrochloride, Tocris

Selective mGlu1 antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material LY 456236 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H15N3O2·HCl
Formula Weight 317.77
Synonym 6-Methoxy-N-(4-methoxyphenyl)-4-quinazolinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

239050

 
r & d systems
239050
50mg Each for $697.40

239010

 
r & d systems
239010
10mg Each for $168.15
Description & Specifications

Specifications

Formula Weight 317.77
Quantity 50mg
Chemical Name or Material LY 456236 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H15N3O2·HCl
CAS 338736-46-2
Synonym 6-Methoxy-N-(4-methoxyphenyl)-4-quinazolinamine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature