Please login to your online account to display your discounted pricing

(±)-LY 395756, Tocris

Mixed mGlu2 agonist/ mGlu3 antagonist

$341.71 - $1,410.00

Chemical Identifiers

Chemical Name or Material (±)-LY 395756
Assay Percent Range >98%
Molecular Formula C9H13NO4
Formula Weight 199.21
Synonym (1S,2S,4R,5R,6S)-rel-2-Amino-4-methylbicyclo[3.1.0]hexane-2,6-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

327210

 
r & d systems
RDS327210
10mg Each for $341.71

327250

 
r & d systems
RDS327250
50mg Each for $1,410.00
Description & Specifications

Specifications

Formula Weight 199.21
Quantity 10mg
Chemical Name or Material (±)-LY 395756
Assay Percent Range >98%
Molecular Formula C9H13NO4
CAS 852679-66-4
Synonym (1S,2S,4R,5R,6S)-rel-2-Amino-4-methylbicyclo[3.1.0]hexane-2,6-dicarboxylic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C