Please login to your online account to display your discounted pricing

LY 393558, Tocris

Dual 5-HT1B/1D antagonist and 5-HT re-uptake inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material LY 393558
Assay Percent Range >98%
Molecular Formula C26H31FN4O4S2
Formula Weight 546.68
Synonym 1-[2-[4-(6-Fluoro-1H-indol-3-yl)-3,6-dihydro-1(2H)-pyridinyl]ethyl]-3,4-dihydro-3-(1-methylethyl)-6-(methylsulfonyl)-1H-2,1,3-benzothiadiazine-2,2-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

335010

 
r & d systems
RDS335010
10mg Each for $257.85

335050

 
r & d systems
RDS335050
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 546.68
Quantity 10mg
Chemical Name or Material LY 393558
Assay Percent Range >98%
Molecular Formula C26H31FN4O4S2
CAS 271780-64-4
Synonym 1-[2-[4-(6-Fluoro-1H-indol-3-yl)-3,6-dihydro-1(2H)-pyridinyl]ethyl]-3,4-dihydro-3-(1-methylethyl)-6-(methylsulfonyl)-1H-2,1,3-benzothiadiazine-2,2-dioxide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C