Please login to your online account to display your discounted pricing

LY 379268, Tocris

Highly selective group II mGlu agonist

$443.80 - $1,848.95

Chemical Identifiers

Chemical Name or Material LY 379268
Assay Percent Range >99%
Molecular Formula C7H9NO5
Formula Weight 187.15
Synonym (1R,4R,5S,6R)-4-Amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

245310

 
r & d systems
RDS245310
10mg Each for $443.80

245350

 
r & d systems
RDS245350
50mg Each for $1,848.95
Description & Specifications

Specifications

Formula Weight 187.15
Quantity 10mg
Chemical Name or Material LY 379268
Assay Percent Range >99%
Recommended Storage Store at +4°C
CAS 191471-52-0
Molecular Formula C7H9NO5
Synonym (1R,4R,5S,6R)-4-Amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
Safety and Handling

Store at +4°C