Please login to your online account to display your discounted pricing

LY 367385, Tocris

Selective mGlu1a antagonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material LY 367385
Assay Percent Range >99%
Molecular Formula C10H11NO4
Formula Weight 209.2
Synonym (S)-(+)-α-Amino-4-carboxy-2-methylbenzeneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

123750

 
r & d systems
RDS123750
50mg Each for $904.70

123710

 
r & d systems
RDS123710
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 209.2
Quantity 50mg
Chemical Name or Material LY 367385
Assay Percent Range >99%
Molecular Formula C10H11NO4
CAS 198419-91-9
Synonym (S)-(+)-α-Amino-4-carboxy-2-methylbenzeneacetic acid
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature