Please login to your online account to display your discounted pricing

LY 364947, Tocris

Selective inhibitor of TGF-βRI

$121.23 - $252.39

Chemical Identifiers

Chemical Name or Material LY 364947
Assay Percent Range >99%
Molecular Formula C17H12N4
Formula Weight 272.31
Synonym 4-[3-(2-Pyridinyl)-1H-pyrazol-4-yl]-quinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

271810

 
r & d systems
RDS271810
10mg Each for $252.39

27181

 
r & d systems
RDS27181
1mg Each for $121.23
Description & Specifications

Specifications

Formula Weight 272.31
Quantity 10mg
CAS 396129-53-6
Synonym 4-[3-(2-Pyridinyl)-1H-pyrazol-4-yl]-quinoline
Assay Percent Range >99%
Chemical Name or Material LY 364947
Molecular Formula C17H12N4
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature