Please login to your online account to display your discounted pricing

LY 354740, Tocris

Potent and highly selective group II mGlu agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material LY 354740
Assay Percent Range >98%
Molecular Formula C8H11NO4
Formula Weight 185.18
Synonym (1S,2S,5R,6S)-2-Aminobicyclo[3.1.0]hexane-2,6-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

324610

 
r & d systems
RDS324610
10mg Each for $252.39

324650

 
r & d systems
RDS324650
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 185.18
Quantity 10mg
Chemical Name or Material LY 354740
Assay Percent Range >98%
Synonym (1S,2S,5R,6S)-2-Aminobicyclo[3.1.0]hexane-2,6-dicarboxylic acid
CAS 176199-48-7
Molecular Formula C8H11NO4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C