Please login to your online account to display your discounted pricing

LY 341495 disodium salt, Tocris™

Disodium salt of LY 341495

$146.74 - $1,307.92

Chemical Identifiers

Chemical Name or Material LY 341495 disodium salt
Assay Percent Range >97%
Molecular Formula C20H17NNa2O5
Formula Weight 397.33
Synonym (2S)-2-Amino-2-[(1S, 2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

40621

 
r & d systems
RDS40621
1mg Each for $146.74

406210

 
r & d systems
RDS406210
10mg Each for $316.20

406250

 
r & d systems
RDS406250
50mg Each for $1,307.92
Description & Specifications

Specifications

Formula Weight 397.33
Quantity 1mg
Assay Percent Range >97%
Synonym (2S)-2-Amino-2-[(1S, 2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid disodium salt
Molecular Formula C20H17NNa2O5
Recommended Storage Store at -20°C
Chemical Name or Material LY 341495 disodium salt
Safety and Handling

Store at -20°C