Please login to your online account to display your discounted pricing

LY 310762 hydrochloride, Tocris

5-HT1D-preferring antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material LY 310762 hydrochloride
Assay Percent Range >98%
Molecular Formula C24H27FN2O2·HCl
Formula Weight 430.94
Synonym 1-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-1, 3-dihydro-3, 3-dimethyl-2H-indol-2-one hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

307810

 
r & d systems
RDS307810
10mg Each for $214.12

307850

 
r & d systems
RDS307850
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 430.94
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material LY 310762 hydrochloride
CAS 192927-92-7
Molecular Formula C24H27FN2O2·HCl
Synonym 1-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-1, 3-dihydro-3, 3-dimethyl-2H-indol-2-one hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature