missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals LY 294002, 98%
$158.23 - $708.95
Chemical Identifiers
CAS | 154447-36-6 |
---|---|
Molecular Formula | C19H17NO3 |
Synonym | 2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one; |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67289LB0
|
Thermo Scientific Chemicals
J67289LB0 |
5 mg |
Each for $158.23
|
|
|||||
AAJ67289MB
|
Thermo Scientific Chemicals
J67289MB |
25 mg |
Each for $708.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
154447-36-6 | |
C19H17NO3 | |
2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one; |
Specifications
5 mg | |
-30°C to -10°C | |
C19H17NO3 | |
2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one; | |
CZQHHVNHHHRRDU-UHFFFAOYSA-N | |
2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one | |
98% |
White to pale yellow | |
154447-36-6 | |
307.35g/mol | |
solid | |
O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1 | |
307.35 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only