Please login to your online account to display your discounted pricing

LY 255283, Tocris

Selective, competitive BLT2 receptor antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material LY 255283
Assay Percent Range >98%
Molecular Formula C19H28N4O3
Formula Weight 360.46
Synonym 1-[5-Ethyl-2-hydroxy-4-[[6-methyl-6-(1H-tetrazol-5-yl)heptyl]oxy]phenyl]ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

220850

 
r & d systems
220850
50mg Each for $924.23

220810

 
r & d systems
220810
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 360.46
Quantity 50mg
Chemical Name or Material LY 255283
Assay Percent Range >98%
Molecular Formula C19H28N4O3
CAS 117690-79-6
Synonym 1-[5-Ethyl-2-hydroxy-4-[[6-methyl-6-(1H-tetrazol-5-yl)heptyl]oxy]phenyl]ethanone
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C