Please login to your online account to display your discounted pricing

LY 2365109 hydrochloride, Tocris

Potent and selective GlyT1 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material LY 2365109 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H27NO5·HCl
Formula Weight 421.91
Synonym N-[2-[4-(1, 3-Benzodioxol-5-yl)-2-(1, 1-dimethylethyl)phenoxy]ethyl]-N-methylglycine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

339710

 
r & d systems
RDS339710
10mg Each for $252.39

339750

 
r & d systems
RDS339750
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 421.91
Quantity 10mg
Chemical Name or Material LY 2365109 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H27NO5·HCl
Synonym N-[2-[4-(1, 3-Benzodioxol-5-yl)-2-(1, 1-dimethylethyl)phenoxy]ethyl]-N-methylglycine hydrochloride
CAS 868265-28-5
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature