Please login to your online account to display your discounted pricing

LY 235959, Tocris


Competitive NMDA antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS101910

 View more versions of this product

Catalog No. 101910

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material LY 235959
CAS 137433-06-8
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C11H20NO5P
Formula Weight 277.26
Quantity 10mg
Synonym [3S-(3α,4aα,6β,8aα)]-Decahydro-6-(phosphonomethyl)-3-isoquinolinecarboxylic acid)

Specifications

Chemical Name or Material LY 235959
Assay Percent Range >98%
Molecular Formula C11H20NO5P
Formula Weight 277.26
Synonym [3S-(3α,4aα,6β,8aα)]-Decahydro-6-(phosphonomethyl)-3-isoquinolinecarboxylic acid)
Accessories

Desiccate at +4°C