Please login to your online account to display your discounted pricing

LY 235959, Tocris

Competitive NMDA antagonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material LY 235959
Assay Percent Range >98%
Molecular Formula C11H20NO5P
Formula Weight 277.26
Synonym [3S-(3α,4aα,6β,8aα)]-Decahydro-6-(phosphonomethyl)-3-isoquinolinecarboxylic acid)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

101910

 
r & d systems
RDS101910
10mg Each for $221.01

101950

 
r & d systems
RDS101950
50mg Each for $904.70
Description & Specifications

Specifications

Chemical Name or Material LY 235959
CAS 137433-06-8
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C11H20NO5P
Formula Weight 277.26
Quantity 10mg
Synonym [3S-(3α,4aα,6β,8aα)]-Decahydro-6-(phosphonomethyl)-3-isoquinolinecarboxylic acid)
Safety and Handling

Desiccate at +4°C