Please login to your online account to display your discounted pricing

LY 215840, Tocris

5-HT2/5-HT7 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material LY 215840
Assay Percent Range >98%
Molecular Formula C24H33N3O2
Formula Weight 395.54
Synonym [8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

152350

 
r & d systems
152350
50mg Each for $976.15

152310

 
r & d systems
152310
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 395.54
Quantity 50mg
Assay Percent Range >98%
Chemical Name or Material LY 215840
CAS 137328-52-0
Recommended Storage Store at +4°C
Molecular Formula C24H33N3O2
Synonym [8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
Safety and Handling

Store at +4°C