missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ly-108 Mouse anti-Mouse, APC, Clone: 13G3-19D, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $355.08

Specifications

Antigen Ly-108
Clone eBio13G3-19D (13G3-19D)
Host Species Murine
Gene Alias SLAM Family Receptor, SLAMF6
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123084 Life Technologies
LS17150880
25μg Each for $143.88
5015090 Life Technologies
LS17150882
100μg Each for $355.08
Specifications

Description

The eBio13G3-19D monoclonal antibody reacts with mouse Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is also known as NTB-A (NK cell, T cell, B cell antigen), and is expressed on NK, T cells, and B cells. Engagement of the Ly108 receptor enhances NK cell activity and expression of IFN-gamma, and contributes to the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. CD4+ T cells from Ly108 knock-out mice exhibit diminished ability to produce IL-4 and defective neutrophil functions.

Staining has not revealed any strain differences between C57BL/6 and BALB/c mice. Other strains have not been tested.
Description & Specifications

Specifications

Ly-108
Murine
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio13G3-19D (13G3-19D)
SLAM Family Receptor, SLAMF6
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL