Please login to your online account to display your discounted pricing

Luzindole, Tocris


Competitive melatonin MT1/MT2 antagonist

Quantity 10mg

Manufacturer: r & d systems  087710

 View more versions of this product

Catalog No. 087710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 292.38
CAS 117946-91-5
Molecular Formula C19H20N2O
Recommended Storage Store at -20°C
Synonym N-Acetyl-2-benzyltryptamine
Chemical Name or Material Luzindole
Assay Percent Range >99%

Specifications

Chemical Name or Material Luzindole
Assay Percent Range >99%
Molecular Formula C19H20N2O
Formula Weight 292.38
Synonym N-Acetyl-2-benzyltryptamine
Accessories

Store at -20°C