missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium powder, -40 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4416703

 View more versions of this product

Catalog No. AA4416703



Description

Description

  • Packaged under Argon
Specifications

Specifications

Lutetium powder
7439-94-3
Lu
UN3089
5h0doz21uj
[Lu]
Odorless
23929
Powder
Air and moisture sensitive
1 g
(REO)
MFCD00011098
−40 Mesh
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutetium
174.967
CHEBI:33382
99.9%
Silver

Chemical Identifiers

7439-94-3
174.967
OHSVLFRHMCKCQY-UHFFFAOYSA-N
23929
lutetium
Lu
MFCD00011098
5h0doz21uj
CHEBI:33382
[Lu]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-103-0

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Argon

Documents