missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium pieces, distilled dendritic, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1241003

 View more versions of this product

Catalog No. AA1241003



Description

Description

  • Packaged under argon
Specifications

Specifications

Lutetium pieces
1g
7439-94-3
(REO)
MFCD00011098
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
[Lu]
174.967
CHEBI:33382
99.9%
Silver
Distilled dendritic
Odorless
Lu
UN3178
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutetium
23929
Pieces

Chemical Identifiers

7439-94-3
174.967
OHSVLFRHMCKCQY-UHFFFAOYSA-N
23929
lutetium
Lu
MFCD00011098
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
CHEBI:33382
[Lu]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents