missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) sulfate octahydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1456603

 View more versions of this product

Catalog No. AA1456603



Specifications

Specifications

Lutetium(III) sulfate octahydrate
99.9%
MFCD00011499
782.24 (683.12 Anhydrous)
Hygroscopic
13473-77-3
(REO)
14,5615
Crystalline
1g

Chemical Identifiers

13473-77-3
MFCD00011499
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents