Please login to your online account to display your discounted pricing

Lutetium(III) oxide, REacton™, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lutetium(III) oxide, REacton™
CAS 12032-20-1
Name Note (REO)
Molecular Formula Lu2O3
Formula Weight 397.94
MDL Number MFCD00011100
Merck Index 14,5615
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1125514

  SDS
alfa aesar
1125514
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 397.94
Quantity 25g
MDL Number MFCD00011100
CAS 12032-20-1
TSCA TSCA
Name Note (REO)
Physical Form Powder
Sensitivity Hygroscopic
Molecular Formula Lu2O3
Odor Odorless
Assay 99.9%
Chemical Name or Material Lutetium(III) oxide, REacton™
Density 9.420
Merck Index 14,5615
Melting Point 2487°C