missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lutetium(III) oxide, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1125506

 View more versions of this product

Catalog No. AA1125506



Specifications

Specifications

Lutetium(III) Oxide
99.9% (REO)
MFCD00011100
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
Powder
Odorless
2487°C
12032-20-1
Lu2O3
White powder
397.94
9.420g/cm3 at 20°C
White
5g

Chemical Identifiers

12032-20-1
Lu2O3
MFCD00011100
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
Safety and Handling

Safety and Handling

EINECSNumber : 234-764-3

Documents