Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) nitrate hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium(III) nitrate hydrate
CAS 100641-16-5
Linear Formula Lu(NO3)3·xH2O
Formula Weight 360.98 (Anhydrous)
MDL Number MFCD00150421
UN Number UN1477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1456803

  SDS
alfa aesar™
1456803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1456806

  SDS
alfa aesar™
1456806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1456814

  SDS
alfa aesar™
1456814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 360.98 (Anhydrous)
Assay 99.9%
Sensitivity Hygroscopic
UN Number UN1477
MDL Number MFCD00150421
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Linear Formula Lu(NO3)3·xH2O
CAS 100641-16-5
Quantity 1g
Health Hazard 1 Danger
TSCA TSCA
Physical Form Crystalline Aggregates
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lutetium(III) nitrate hydrate
Assay Percent Notes (REO)
Color White
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.