Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) iodide, ultra dry, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium(III) iodide
CAS 13813-45-1
Name Note Ultra dry
Formula Weight 555.68
MDL Number MFCD00049970
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3580303

  SDS
alfa aesar™
3580303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3580306

  SDS
alfa aesar™
3580306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Flakes or Powder
Formula Weight 555.68
Packaging Ampule
TSCA TSCA
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 5.60g/mL
Sensitivity Hygroscopic
Storage Note 1
  • Ampouled under argon
Melting Point 1050°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Ultra dry
Assay Percent Notes (REO)
Health Hazard 1 Warning
MDL Number MFCD00049970
Chemical Name or Material Lutetium(III) iodide
Assay 99.9%
CAS 13813-45-1

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon